Order Entry
Austria
ContactUsLinkComponent
 
Tin Standard Solution, 1000 mg/L Sn in Diluted HNO₃ and HF for ICP-MS
  ACSDM255.1N05FP.L1
 :  CPACHEM
 :  
Tin Standard Solution, 1000 mg/L Sn in Diluted HNO₃ and HF for ICP-MS
  ACSDM255.1N05FP.L1
 :  CPACHEM
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    1% HNO₃/0,5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Tin
  • Symbol:
    Sn