Order Entry
Austria
ContactUsLinkComponent
 
Rhodium Standard Solution, 1000 mg/L Rh in Diluted Hydrochloric Acid for ICP-MS
  ACSDM247.2CP.L1
 :  CPACHEM
 :  
Rhodium Standard Solution, 1000 mg/L Rh in Diluted Hydrochloric Acid for ICP-MS
  ACSDM247.2CP.L1
 :  CPACHEM
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Rhodium
  • Symbol:
    Rh