Order Entry
Austria
ContactUsLinkComponent
Platinum Standard Solution, 1000 mg/L Pt in Diluted Hydrochloric Acid for ICP-MS
Platinum Standard Solution, 1000 mg/L Pt in Diluted Hydrochloric Acid for ICP-MS
Catalog # ACSDM244.2CP.L1
Supplier:  CPACHEM
CAS Number:  
Platinum Standard Solution, 1000 mg/L Pt in Diluted Hydrochloric Acid for ICP-MS
Catalog # ACSDM244.2CP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Platinum
  • Symbol:
    Pt

Specifications

About this item