Order Entry
Austria
ContactUsLinkComponent
Lithium Standard Solution, 1000 mg/L Li in Diluted Nitric Acid for ICP-MS
Lithium Standard Solution, 1000 mg/L Li in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM230.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Lithium Standard Solution, 1000 mg/L Li in Diluted Nitric Acid for ICP-MS
Catalog # ACSDM230.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item