Order Entry
Austria
ContactUsLinkComponent
 
Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
  ACSDM258.2NP.L1
 :  CPACHEM
 :  
Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
  ACSDM258.2NP.L1
 :  CPACHEM
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Terbium
  • Symbol:
    Tb

 

 

Tb 1000 mg/L in diluted nitric acid for ICP/MS.

Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.

 : ISO 17034, ISO 17025, ISO 9001.