Order Entry
Austria
Orders LinkContactUsLinkComponent
 

High Precision Reference Thermometer, P795

P795, −200...+850 °C, ±0,015 °C from −100...+199,99 °C
±0,025% remaining range, 200×93×44 mm, 600 g
  DOST5000-0795
High Precision Reference Thermometer, P795
  DOST5000-0795
High Precision Reference Thermometer, P795

P795, −200...+850 °C, ±0,015 °C from −100...+199,99 °C
±0,025% remaining range, 200×93×44 mm, 600 g

 
1 560,00 €
   
1 * 1 ST
 

An instrument which meets the demands. The 2-channel-resistance thermometer automatically detects our Pt100-smartprobes with integrated EEprom. Thereby the calibration characteristic of each probe is transferred automatically to the instrument.

  • 2-channel-instruments, for PT100 according EN 60751
  • Simultaneous display of two measured values or differential measurement
  • Recording maximum, minimum, hold and average values (average over user selectable time)
  • Integrated calibration function (acc. EN60751 up to 14 points)
  • High measuring accuracy (+0,010 °C)
  • Includes USB-cable
  • Data Logger function for up to 6000 measurements
  • Optional DAKKS or ISO-system calibration certification

The thermometer will achieve a system accuracy of ±0,015 K (instrument + probe). Both measuring channels can be easily seen on the large LC display. The instrument contains numerous functions for the measurement visualisation. Various integrated calibration functions, including the intelligent EEprom-probes, takes this compact instrument to a high performance class, making it ideally suitable as a primary reference standard.

 
  • Model:
    P795
  • Range:
    −200...+850 °C
  • Weight:
    600 g
  • W×D×H:
    200×93×44 mm
  • Accuracy:
    ±0,015 °C from −100...+199,99 °C
    ±0,025% remaining range
  • Cat. No.:
    DOST5000-0795
  • No. of channels:
    2
  • Memory:
    ~6000 measurements
  • Housing material:
    ABS
  • Display:
    2 line LCD
  • Operating temperature:
    0...40 °C
Accessories

 

  •  
    Description
     
     
  • DOST5990-0001
    Block battery, 9 V
       
    1 * 1 ST - 5,70 €