ContactUsLinkComponent
Hei-VAP Industrial - Performance Plus Package Rotary Evaporators
Hei-VAP Industrial - Performance Plus Package Rotary Evaporators
Catalog # HEID518-46100-00
undefined
Hei-VAP Industrial - Performance Plus Package Rotary Evaporators
Catalog # HEID518-46100-00
Specifications
  • Description:
    Hei-VAP industrial - performance plus package
  • Pk:
    1
Specifications
About this item
Hei-VAP industrial - performance plus package.
  • This packages includes:
  • Hei-VAP industrial model with glassware A1C - For low-boiling solvents and highest recovery rates
  • Base cart
  • Tubing set with 16 meter piece incl. hose clamps
  • Vacuum pump Hei-VAC valve industrial
  • Chiller Hei-CHILL 5000
  • 40 L - Kryo 30 thermal fluid