Tetradecyltrimethylammonium bromide BP, Ph.Eur., pure, Pharma Grade
Catalog # APPC142542.1209
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:200 g
- Seq:3505
Specifications
Specification Test Results
- Solubility:Water 500 g/L at 20 °C
- Appearance of solution:Passes test
- Amines and amine salts:Passes test
- Acidity or alkalinity:Passes test
- Insoluble matter in H₂O:Passes test
- Loss on drying at 105 °C:2,0%
- Residue on ignition (as SO₄):0,5 %
Specification Test Results
About this item
:
:336.394 g/mol
UN:1759
ADR:8,II
:GHS05
CAS Number:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770