ESD Safety Ankle Boots, Lace-Up, Quentin Mid S3 S
: ELTEN
ESD safety ankle boot made from hydrophobised suede/microfibre upper and a PU/PU sole (Crossworker).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO black with recycled material content
- Non-metallic, penetration-resistant midsole
- Steel toe cap
- PU scuff cap
Colour: Black/blue
: EN ISO 20345:2022 S3S FO/SR/SC/LG, form B.
- Catalog No:
- ELTN763811/46
- ELTN763811/45
- ELTN763811/44
- ELTN763811/48
- ELTN763811/47
- ELTN763811/43
- ELTN763811/42
- ELTN763811/41
- ELTN763811/40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together