Order Entry
Austria
ContactUsLinkComponent
 
1,5-Diaminopentane
Catalog # CMPLSB65012.250MG
CAS Number:  
1,5-Diaminopentane
Catalog # CMPLSB65012.250MG
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :102.18 g/mol
Boiling Pt:178...180 °C
Melting Pt:9 °C
Density:0,873 g/cm³
Flash Pt:62 °C
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:462-94-2
Formula:C₅H₁₄N₂
MDL Number:MFCD00008239