Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
pH Calibration Buffers
pH Buffer
pH Buffer
pH Buffer
CMPLPH112.1L
:
CUSTOM MADE CHEMICALS LAB
:
pH Buffer
CMPLPH112.1L
:
CUSTOM MADE CHEMICALS LAB
:
Pk:
1 L
Description:
di-Sodium hydrogen phosphate/Sodium hydroxide
pH accuracy:
12,0 at 20 °C
Final volume:
1000 ml
:
ISO 9001.
:
Store under normal laboratory conditions, at temperatures between 15 to 25 °C.