EPA Method 8082A Surrogate Standard
Catalog # EHERA50000424IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Concentration:1000 µg/ml in isooctane
- Pk:1 mL
- Description:EPA method 8082A surrogate standard mixture 424
- Type:Native/Parent
Specifications
About this item
Caution: Laboratory use.