- Packed:200 combo packets/case
- Colour:White
- Description:M-Zone ScreenWipes™
- L×W:22,9×20,3 cm (9×8")
- Pk:200
A combo pack of two individually packaged Sontara® fabric wipes; one dry and one pre-saturated with Lab & Glass Clean and a minimal amount of fast-drying IPA to clean displays and touchscreens.
- Low-linting Sontara® fabric
- One dry wipe and one pre-saturated wipe with Lab & Glass Clean and IPA
- Convenient for equipment monitor, display, and touchscreen cleaning
- Good compatibility with common disinfectants
- Polycellulose blend wipes are 22,9×20,3cm folded in 12,7×7,6cm packs
- Not irradiated
Designed for use in cleanrooms and controlled environments, M-Zone ScreenWipes™ can be used on monitors, touchscreens, lenses, and safety glasses. Made from low-linting Sontara® fabric impregnated with highly filtered Lab & Glass Clean and a minimal amount of IPA, M-Zone ScreenWipes™ have considerably less VOC content and are a gentler alternative to the harsh ammonia-based screen and instrument wipes available to the industry. With a one/two action, the wet wipe cleans and removes streaks and contaminants without oversaturating the surface to be cleaned. The dry wipe buffs the surface to a streak-free finish. These wipes specifically address screen and lens cleaning applications in aerospace, hi-tech manufacturing, and clean production.
Part number and lot number printed on each case. Certificate of Conformance available upon request.