Order Entry
Austria
ContactUsLinkComponent
Pharmaceutical Analytical Standard, Biotin for System Suitability, Reference Material
Pharmaceutical Analytical Standard, Biotin for System Suitability, Reference Material
Catalog # EDQMY0001978
Supplier:  EDQM
CAS Number:  
Pharmaceutical Analytical Standard, Biotin for System Suitability, Reference Material
Catalog # EDQMY0001978
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical reference standard
  • CAS No.:
    58-85-5
  • Description:
    Biotin for system suitability
  • MDL No.:
    MFCD00005541
  • Synonym(s):
    Biotin, D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H
  • Melting point:
    231...233 °C (lit.)
  • Molecular weight:
    244,31
  • Storage temperature:
    −20 °C
  • Form:
    Solid

Specifications

About this item

Biotin is a hydro-soluble vitamin, which helps in the health of skin, nerve, digestive system and also serves as an important cofactor for mammalian carboxylases. It is found within the cells in free and bound pools and assists in releasing energy and metabolism of fats, protein and carbohydrates.

Biotin may be used as an EP reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography technique.