ContactUsLinkComponent
Liquid Amies, Sterile, Transporter Swab
  DELT300284.SE
 :  DELTALAB
Liquid Amies, Sterile, Transporter Swab
  DELT300284.SE
 :  DELTALAB
  • Sterile:
    +
  • Description:
    Shaft material: snappable PS, tip material: viscose
  • Transport medium:
    Amies liquid
  • Vial capacity:
    0,0550 m³
  • Pk:
    1.200
This is a variation of the traditional Amies media swab where the media is presented on liquid form without agar.
  • Peel-pack dimensions: 38×210 mm
Media is supplied in its liquid form, in a sponge, suitable for direct extensions on slides.
Swabs are sterilised by radiation.
Suitable for conservation of pathogens such as:Haemophilus sp., Corynebacterium sp., Streptococcus pyogenes, Streptococcus pneumoniae, Shigella flexneri, Salmonella typhi, Brucella abortus, Staphylococcus epidermidis, Escherichia coli.
 : CE MDD Class IIa. CE IVD.
 : Expiry date: 30 months after sterilisation date.