Phosphate Buffer Solution
Phosphate buffer solution for in vitro diagnostic use, 1
Catalog # 1805-1VL
Supplier: SIGMA ALDRICH MICROSCOPY
Jump to:
Product Details & Documents
Contains sodium and potassium phosphates. Intended for use in the Sigma Butyrate Esterase Procedure, No. 181B.
Dokumente
Merck The Color of Tissue Diagnostics
Produktliteratur
Specifications
Specifications
- Pk:1
- Description:Phosphate buffer solution for in vitro diagnostic use
- Cat. No.:1805-1VL
- Form:Powder
- pH:7,60 - 7,80 (25 °C)
- Storage temperature:RT



