Orders LinkContactUsLinkComponent
Magnifier lamps, varioLED flex
Supplier:   ESCHENBACH OPTIK
Magnifier lamps, varioLED flex
Supplier:  ESCHENBACH OPTIK
Certificates
Magnifier lamps, varioLED flex
Magnifier lamps, varioLED flex
2 Options Available Below
Product Details & Documents
These illuminated bench magnifiers are suitable for use in the scientific and professional fields, particularly for detailed technical work and reading.
  • Aspheric PXM® lightweight lens, cera-tec® coated for a distortion-free image with edge-to-edge sharpness
  • Segment switching provides a choice between ring illumination and segment illumination for closer inspection of different surface structures
  • Illumination level: approx. 6000 lx (at a working distance of 150 mm)
  • Gooseneck design with protective cover for improved hygiene
Low power consumption as power requirement is only 5 W, service life of LED approx. 50000 hours.
Product Family Options
Product Information
  • Magnification
    Ø
    Availability
    Price
  • 630-2154
    Magnifier lamp, 350 mm
    Magnifier lamp, 350 mm
    2,5×
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
  • 630-2155
    Magnifier lamp, 600 mm
    Magnifier lamp, 600 mm
    2,5×
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 805,00 €
    805,00 €
    1 * 1 ST
       805,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Table base, steel with black finish (2 kg) for varioLED flex magnifier lamps
    Table base, steel with black finish (2 kg) for varioLED flex magnifier lamps
    630-2156
    Log in for availability
    1 * 1 ST - 285,00 €