Order Entry
Austria
ContactUsLinkComponent
Peroxidase Indicator Reagent
Peroxidase Indicator Reagent
Catalog # 3901-10VL
CAS Number:  
Peroxidase Indicator Reagent
Catalog # 3901-10VL
CAS Number:  

Specifications

  • Pk:
    10
  • Form:
    Powder
  • Shipping conditions:
    Wet ice
  • pH range:
    7,0 (20 °C)
  • Storage temperature:
    2...8 °C
  • Application:
    Hematology and histology, for in vitro diagnostic use

Specifications

About this item

It contains p-Phenylenediamine diHCL (1 part) and catechol (2 parts). Commonly referred to as Hanker Yates reagent. Kit component in the Sigma Leukocyte peroxidase (myeloperoxidase) Procedure, 390A.