Express Eingabe
Austria
AuftragsverfolgungKontakt
Multi-Parameter Tester, CombiCheck 80
CombiCheck 80 Muliti-parameter standard solutions, 1 KIT
Catalog # 713-4103
Supplier:   WTW
Multi-Parameter Tester, CombiCheck 80
Catalog # 713-4103
Supplier:  WTW
CAS Number:  
Multi-Parameter Tester, CombiCheck 80
Multi-Parameter Tester, CombiCheck 80
CombiCheck 80 Muliti-parameter standard solutions, 1 KIT
CAS Number:  
Price
166,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
CombiCheck solutions are ready-to-use multi-parameter standards, Each package contains a standard solution and an addition solution. Both solutions can be used directly without dilution for quality assurance.
  • The standard solution is used to check the accuracy of the results for the complete system: procedure - analytical method - reagents - photometer
  • The addition solution is used to check sample-dependent influences by measuring the recovery rate and to determine the most suitable sample preparation method
The maximum number of determinations which can be made with a CombiCheck standard solution depends on the test set used. With the addition solution 280 determinations are always possible.
Parameters can be checked with their concentrations as follows:
COD: 1,500 mg/L COD
Nitrate: 25,0 mg/L NO₃-N
Phosphate: 15,0 mg/L PO₄-P
Specifications
Specifications
  • VE:
    1 KIT
  • Bezeichnung:
    CombiCheck 80 Muliti-parameter standard solutions
Specifications
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.