Order Entry
Austria
ContactUsLinkComponent
Memo Rail Transmitters
Memo Rail Transmitters
Catalog # KNICA1401N-P1-21
Supplier:  KNICK
Memo Rail Transmitters
Catalog # KNICA1401N-P1-21
Supplier:  KNICK

Specifications

  • Output:
    4 - 20 mA
  • Description:
    Memo Rail A1401N-P1-214-wire
  • Power:
    24 V DC; 90 - 230 V AC
  • Pk:
    1
  • Tube Ø:
    12,5 mm

Specifications

About this item

The Knick A1401N-P1-21, stands out in the market for its unparalleled performance and reliability.

  • Superior Quality
  • Cost-Effective
  • Fast Delivery

Memo Rail A1401N-P1-214-wire DIN for the determination of pH, Conductivity or dissolved oxygen with Memosens sensors measurement range can be selected via DIP switches2 with plug-P1screw.