0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Cationic Standards
Silver Standards
Silver Standard Solution
Silver Standard Solution
Catalog # 713-4093
Supplier:
WTW
CAS Number:
Expand image
Silver Standard Solution
Catalog # 713-4093
Supplier:
WTW
CAS Number:
Specifications
Concentration:
1000 mg/L
Pk:
500 mL
Description:
Silver standard solution
Specifications
About this item
Product Details
Product Details