Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Cadmium Standards
Cadmium Standard Solution
Cadmium Standard Solution
713-4076
:
WTW
:
Cadmium Standard Solution
713-4076
:
WTW
:
Concentration:
1000 mg/L
Pk:
500 mL
Description:
Cadmium standard solution