Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Boron Standards
Boron Standard Solution
Boron Standard Solution
Boron Standard Solution
Catalog # 713-4075
Supplier:
WTW
CAS Number:
Expand image
Boron Standard Solution
Catalog # 713-4075
Supplier:
WTW
CAS Number:
Specifications
Concentration:
1000 mg/L
Pk:
500 mL
Description:
Boron standard solution
Specifications
About this item
Product Details
Product Details