ContactUsLinkComponent
Analog Conductivity Measuring Cells for Wastewater
 :  WTW
Analog Conductivity Measuring Cells for Wastewater
 :  WTW
  • Catalog No:
  • WTWA302222
  • WTWA302220
  • WTWA302225
  • Sensor type:
  • Integrated NTC
  • Integrated NTC
  • Integrated NTC
  • Conductivity range:
  • 0,001...200 µS/cm
  • 0,001...200 µS/cm
  • 1 µS/cm...2 S/cm
  • Cell constant:
  • 0,1 cm⁻¹, ±2%
  • 0,1 cm⁻¹, ±2%
  • 0,475 cm⁻¹, ±1,5%
Frequently Bought Together