Order Entry
Austria
Orders LinkContactUsLinkComponent
Mounting Medium, CC/Mount™

Mounting Medium, CC/Mount™

Aqueous-based permanent mounting medium, 30 mL
Catalog # C9368-30ML
Mounting Medium, CC/Mount™
Catalog # C9368-30ML
CAS Number:  
Mounting Medium, CC/Mount™

Aqueous-based permanent mounting medium, 30 mL

CAS Number:  
Price
110,00 €
Log in for availability
1 * 30 mL
Product Details & Documents

CC/mount is an aqueous based permanent mounting medium with a very high refractive index, which when applied to the stained tissue sections can store the tissue specimens permanently without the fading of the chromogens. Because of the superior refractive index of CC/mount, tissues mounted in this medium look like specimens mounted with organic based mounting media. No coverslipping is required with CC/mount. However, if coverslipping is required, dried CC/mount can be post mounted by using an organic based mounting medium. CC/mount permits the long-term storage of antigens localised using organic insoluble chromogen substrates, including AEC, DAB, Fast Red, BCIP/NBT, BCIP/INT and fluorescent dyes like FITC and phycobiliproteins. The high pH of CC/mount ensures increased stability of fluorescence intensity.

  • No need of coverslip
  • No exposure to organic fumes
  • Permanent storage of slides
  • High resolution of tissue specimens

Remove the red end cap from the bottle and cut the tip off with a sharp razor or scissors. This will allow a smooth flow of CC/mount and prevent formation of tiny bubbles because of squeezing.

CC/mount is an aqueous based permanent mounting medium, similar to Crystal Mount™, for the permanent preservation of tissue sections one, two and three stained with peroxidase and alkaline phosphatase based systems as well as with various fluorescent dyes.

Ordering information: Store the product at room temperature. Do not refrigerate.

Documents

Specifications

Specifications

  • Pk:
    30 mL
  • Description:
    Aqueous-based permanent mounting medium
  • Application:
    Hematology, histology

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.