Air Flow Sensors with Integrated Amplifier, LC 521
Supplier: EGE ELKTRONIK SPEZIAL SENSOREN
Certificates
About this item
Air flow sensors with integrated amplifier for explosive areas.
- Low monitoring of gaseous media in hazardous areas
- Installation in zone 2 (gas) and zone 22 (dust)
- Switching point adjustable on site during operation
- Stainless steel housing
Sensor length: 101 mm
Output function: Non linear
Supply voltage: 24 VDC +/- 10%
Current consumption: max. 35 mA
Ex area uf use: Zone 2, Zone 22
Temperature range: –10 to +60 °C
Start-up time: 20 mins
Reaction time: 5 mins
Compressive strength: 1 bar
Display flow: Two-colour-illuminated dot red/green
Electrical connection: 2 m PUR-cable 3×0,5 mm²
Certifications: Certified of Protection [EN 60529] IP 67, ATEX marking gas - Ex II 3 G Ex ic mc IIC T4...T3 Gc, Certificate of conformity - EGE 21.0010 X, Ex Marking ATEX Dust- Ex II 3 D Ex ic mc IIIC T135 °C Dc.
Frequently Bought Together