Order Entry
Austria
ContactUsLinkComponent
Methyl isothiocyanate, Sigma-Aldrich®
Toxic
Corrosive
Environment
Methyl isothiocyanate, Sigma-Aldrich®
Catalog # CMPLSB27280.100MG
CAS Number:  
Toxic
Corrosive
Environment
Methyl isothiocyanate, Sigma-Aldrich®
Catalog # CMPLSB27280.100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Colour):
    White to Yellow to Orange
  • Appearance (Form):
    Solid
  • Infrared spectrum:
    Conforms to Structure
  • Purity (GC):
    ≥96.5%

Specification Test Results

About this item

 :
 :73.12 g/mol
Merck Index:12,06165
Boiling Pt:117 °C (1013 hPa)
Melting Pt:31…37 °C
Density:1.069 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:209-132-5
UN:2477
ADR:6.1,I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS09
CAS Number:556-61-6
Formula:C₂H₃NS
MDL Number:MFCD00004818