Order Entry
Austria
ContactUsLinkComponent
Anti-SEZ6 Rabbit Polyclonal Antibody
Anti-SEZ6 Rabbit Polyclonal Antibody
Catalog # ORIGTA337738
Supplier:  OriGene
Anti-SEZ6 Rabbit Polyclonal Antibody
Catalog # ORIGTA337738
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SEZ6
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    PBS buffer with 0,09% (w/v) sodium azide and 2% sucrose
  • Molecular weight:
    105 kDa
  • Sequence:
    RAPKCLLEQLKPCHGLSAPENGARSPEKQLHPAGATIHFSCAPGYVLKGQ
  • Immunogen:
    The immunogen for anti-SEZ6 antibody is: synthetic peptide directed towards the C-terminal region of Human SEZ6. Synthetic peptide located within the following region: RAPKCLLEQLKPCHGLSAPENGARSPEKQLHPAGATIHFSCAPGYVLKGQ
  • Purification:
    Affinity Purified
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: SEZ6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human