Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Filters
Tangential Flow Cassettes
Omega™ Membrane, Centramate Cassettes
Omega™ Membrane, Centramate Cassettes
Catalog # 516-8831
Supplier:
Cytiva (Formerly Pall Lab)
Expand image
Omega™ Membrane, Centramate Cassettes
Catalog # 516-8831
Supplier:
Cytiva (Formerly Pall Lab)
Specifications
Filtration area:
4600 cm²
MWCO:
300k
Description:
Suspended screen
Seq:
02
Pk:
1
Specifications
About this item
Product Details
Product Details