ContactUsLinkComponent
Disposable Specimen Cuvettes for UNISTAT Bilirubinometers
Disposable Specimen Cuvettes for UNISTAT Bilirubinometers
Catalog # REIC13102140
Supplier:  REICHERT
Disposable Specimen Cuvettes for UNISTAT Bilirubinometers
Catalog # REIC13102140
Supplier:  REICHERT
Specifications
  • Description:
    Disposable specimen cuvettes for UNISTAT bilirubinometers
  • Seq:
    01
  • Pk:
    51
Specifications
About this item
Disposable specimen cuvettes specifically designed for UNISTAT Bilirubinometers. These cuvettes hold 20 μl (microliters) of serum and are placed into the optical path of our Bilirubinometer to measure newborn Total Serum Bilirubin concentration.
Packaging: 51 disposable cuvettes are supplied in a package.