Order Entry
Austria
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99.0% (by GC), pellets suitable for electrophoresis, for molecular biology dust-free, Supelco®
Sodium dodecyl sulphate (SDS) ≥99.0% (by GC), pellets suitable for electrophoresis, for molecular biology dust-free, Supelco®
Artikelnr. SUPL74255-250G
Supplier:  Merck
CAS Number:  
Sodium dodecyl sulphate (SDS) ≥99.0% (by GC), pellets suitable for electrophoresis, for molecular biology dust-free, Supelco®
Artikelnr. SUPL74255-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :288.38 g/mol
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175