Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate ≥99.99% (trace metals basis), Supelco®
Ammonium thiocyanate ≥99.99% (trace metals basis), Supelco®
Catalog # 431354-50G
Supplier:  Merck
CAS Number:  
Ammonium thiocyanate ≥99.99% (trace metals basis), Supelco®
Catalog # 431354-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :76.12 g/mol
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428