Order Entry
Austria
ContactUsLinkComponent
Silica gel pore size 60 Å, 5-25 μm particle size, without binder, high purity suitable for thin layer chromatography (TLC), Supelco®
Silica gel pore size 60 Å, 5-25 μm particle size, without binder, high purity suitable for thin layer chromatography (TLC), Supelco®
Catalog # 288519-250G
Supplier:  Merck
CAS Number:  
Silica gel pore size 60 Å, 5-25 μm particle size, without binder, high purity suitable for thin layer chromatography (TLC), Supelco®
Catalog # 288519-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:> 1600 °C
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232