Order Entry
Austria
ContactUsLinkComponent
Electrolyte solution nonaqueous, 2 M LiCl in ethanol, Supelco®
Electrolyte solution nonaqueous, 2 M LiCl in ethanol, Supelco®
Catalog # SUPL56754-250ML
Supplier:  Merck
CAS Number:  
undefined
Electrolyte solution nonaqueous, 2 M LiCl in ethanol, Supelco®
Catalog # SUPL56754-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Electrolyte solution nonaqueous, 2 M LiCl in ethanol
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    +2°C to +30°C

Specifications

About this item

Filling solution for reference electrodes used for titrations in nonaqueous solutions.