Express Eingabe
Austria
ContactUsLinkComponent
Natriumhydroxid 21% in wässriger Lösung, Supelco®
Corrosive
Natriumhydroxid 21% in wässriger Lösung, Supelco®
Supplier:  Merck
Certificates
Corrosive
Natriumhydroxid 21% in wässriger Lösung, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.219 g/cm3 (20 °C)
  • pH:
    11 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together