Polyvinyl alcohol mounting medium with DABCO®, antifading, Supelco®
Catalog # SUPL10981-100ML
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Description:Polyvinyl alcohol mounting medium with DABCO®, antifading
- M-Clarity quality level:MQ100
- Refractive index:n20/D 1.377
- Storage temperature:2-8°C
- Density:1.08 g/mL at 20 °C
Specifications
About this item
Aqueous solution containing tris(hydroxymethyl)aminomethane, tris(hydroxymethyl)aminomethane hydrochloride, polyvinyl alcohol 22000, glycerol, and 1,4-diazabicyclo[2.2.2]octane.
DABCO is a registered trademark of Air Products & Chemicals, Inc.