Order Entry
Austria
ContactUsLinkComponent
 
 
 
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, Supelco®
  SUPL01709-500ML
 :  Merck
 :  
 
 
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, Supelco®
  SUPL01709-500ML
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

 :
 :6.1,III
 :M-Clarity™ quality level = MQ100
 :3426
 :GHS06
 :GHS08
 :Refrigerator
 :79-06-1
 :MFCD00080848