Orders LinkContactUsLinkComponent
Maxitips, Pipette Tips
Maxitips, Pipette Tips
   Sustainable Options Available
Supplier:   EPPENDORF
Maxitips, Pipette Tips
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Maxitips, Pipette Tips
3 Options Available Below
Product Details & Documents
Eppendorf Maxitips are designed for large volume liquid handling, offering high precision and compatibility with Eppendorf pipetting systems. Ideal for diverse lab applications, they ensure accuracy and ease of use, enhancing workflow efficiency.
  • Use Maxitips P to aspirate 1 to 10 ml from beakers using the positive displacement principle
  • Use Maxitips S to aspirate between 2,5 and 10 ml from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips ensures accurate dispensing of non-aqueous liquids
Experience exceptional levels of flexibility with different Maxipettor pipette tips that allow liquid removal from a variety of vessels. When aspirating from small vessels such as beakers, select the Eppendorf Maxitip P for precise aspiration of 1 to 10ml volumes. For tall, narrow-necked vessels and measuring flasks, use the longer-tip Maxitip S to aspirate volumes of 2,5 to 10 ml.
Designed for use with Maxipettor large volume pipette models from Eppendorf, the Maxitips pipette tips are perfect for all large vessel types.
Ordering information: Please note: Maxitips are designed for use with Eppendorf Maxipettor® 4720 only.
Specifications for Product Family
Specifications
  • Catalog No:
  • 612-7321
  • 612-7333
  • 612-7331
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Product Information
  • Tip style
    Volume
    Packed
    Availability
    Price
  • Maxitips P
    1 - 10 ml
    100 pieces
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 132,00 €
    132,00 €
    1 * 100 ST
       132,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-7321
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Maxitips S
    2,5 - 10 ml
    200 pieces
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 94,60 €
    94,60 €
    1 * 200 ST
       94,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-7333
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Maxitips S starter kit
    2,5 - 10 ml
    100 Maxitips, 10 dispensing Parts, 10 valves
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 99,80 €
    99,80 €
    1 * 1 KIT
       99,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-7331
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact