ContactUsLinkComponent
illustra™ TempliPhi™ sequence resolver kit
illustra™ TempliPhi™ sequence resolver kit
Supplier:  Cytiva
Certificates
undefined
illustra™ TempliPhi™ sequence resolver kit
Supplier:  Cytiva
Specifications
  • Storage conditions:
    Enzyme mix to be stored at -70°C; all other components at -20°C.
    Thaw components on ice at 0 to 4°C during handling