Express Eingabe
Austria
ContactUsLinkComponent
Dye terminator removal, illustra™ AutoSeq™ G-50 columns
Dye terminator removal, illustra™ AutoSeq™ G-50 columns
Supplier:  Cytiva
Certificates
undefined
Dye terminator removal, illustra™ AutoSeq™ G-50 columns
Supplier:  Cytiva
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Bezeichnung:
    illustra™ AutoSeq™

Specifications

Related Information

Frequently Bought Together