illustra™ MicroSpin™ G-25 columns
27-5325-01
: Cytiva
:
- Pk:50
- Application:
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:illustra™ MicroSpin™ G-25 columns
- Environmentally Preferable:
- Description:illustra™ MicroSpin™
For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 to 100 μl using spin column chromatography.
- Prepacked with Sephadex™ G-25 DNA Grade and pre-equilibrated in distilled water containing 0,05% Kathon CG/ICP Biocide
- Ready to use: requires less than 4 minutes from sample application to collection of purified product
- Tested in nickase, single and double-stranded exonuclease and RNase assays
- Can also be used for desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labelled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 μl