Polypropylene glycol, M.W. 2,000
297776T
:
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
- Identification by IR:Passes test
- Colouration:Max. 20 APHA
- Hydroxyl value:53 - 58
- Viscosity (25°C):300 - 340 mPa.s
- Water:Max. 0.05 %
: Poly(propylene glycol), Poly(propylene oxide), PPG
:
:1025
:1.005 g/cm³ (20 °C)
:112.8 °C (closed cup)
:Ambient
:500-039-8
:25322-69-4
:H(OCH₂CH₂CH₂)nOH
:MFCD00084446