Polypropylene glycol, M.W. 2,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
CAS Number:25322-69-4
:
Formula:H(OCH₂CH₂CH₂)nOH
MDL Number:MFCD00084446
Density:1.005 g/cm³ (20 °C)
Molecular Weight:1025
Flash Pt:112.8 °C (closed cup)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-039-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together