Order Entry
Austria
ContactUsLinkComponent
 
Disodium hydrogen phosphate, anhydrous ≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
 :  Avantor
 
undefined
Disodium hydrogen phosphate, anhydrous ≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (Na₂HPO₄):
    ≥99.0 %
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • pH of 5% Solution at 25°C:
    8.7 - 9.3
  • Insoluble Matter:
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Loss on Drying at 105°C:
    ≤0.2 %

 

 

Frequently Bought Together