Order Entry
Austria
ContactUsLinkComponent
Dimethyl disulphide
Flammable
Toxic
Environment
Dimethyl disulphide
Catalog # EHERCA12726480
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Toxic
Environment
Dimethyl disulphide
Catalog # EHERCA12726480
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :94.2 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–98 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:210-871-0
UN:2381
ADR:3,II
 :GHS02
 :GHS06
 :GHS09
CAS Number:624-92-0
Formula:C₂H₆S₂
MDL Number:MFCD00008561