Calcium chloride 0.01 mol/l (0.02 N) in aqueous solution Reag. Ph. Eur. 1014602
Catalog # 87801.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903