(±)-1,2-Octanediol 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
CAS Number:1117-86-8
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00010738
Boiling Pt:243
Molecular Weight:146.23 g/mol
Melting Pt:28…31 °C
Flash Pt:138 °C
EINECS:214-254-7
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together