Order Entry
Austria
ContactUsLinkComponent
Dehydrated Culture Media, Columbia Agar Base
Dehydrated Culture Media, Columbia Agar Base
Catalog # LIOF6100135
Supplier:  LIOFILCHEM
CAS Number:  
Dehydrated Culture Media, Columbia Agar Base
Catalog # LIOF6100135
Supplier:  LIOFILCHEM
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Description:
    Columbia agar base
  • Storage temperature:
    10...30 °C
  • Incubation:
    36 ±1 °C for 18 - 48 hours at 5 - 10% CO₂
  • pH:
    7,3 (±0,2) at 25 °C

Specifications

About this item

Columbia agar base, enriched with sterile sheep blood (5%), is suitable for isolation and growth of fastidious microorganisms such as streptococci, staphylococci, pneumococci and listeriae from clinical samples.