Order Entry
Austria
Orders LinkContactUsLinkComponent
BIOFLOAT™ FLEX Coating Solution | Avantor

BIOFLOAT™ FLEX Coating Solution

BIOFLOAT™ FLEX coating solution, Clear, 60 ml, 1
Catalog # 734-3632
Supplier:   FACELLITATE
BIOFLOAT™ FLEX Coating Solution
Catalog # 734-3632
Supplier:  FACELLITATE
CAS Number:  
BIOFLOAT™ FLEX Coating Solution
BIOFLOAT™ FLEX Coating Solution

BIOFLOAT™ FLEX coating solution, Clear, 60 ml, 1

CAS Number:  
Price
107,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The BIOFLOAT™ FLEX coating solution guarantees the creation of surfaces without unspecific protein or cell binding.

  • Easy and fast procedure
  • Flexibility
  • Robustness
  • Xeno-free and chemically defined
  • Wide range of applications

Self-coating solution for plastic and glass consumables. The BIOFLOAT™ Flex Coating Solution outperforms existing products by its anti-adhesive properties.

Certifications: The product does not use any raw materials of animal or biological origin and therefore does not have TSE/BSE.

Packaging: One bottle per pack.

Caution: Research use only. Not intended for diagnostic or therapeutic use.

Specifications

Specifications

  • Pk:
    1
  • Colour:
    Clear
  • Description:
    BIOFLOAT™ FLEX coating solution
  • Size:
    60 ml
  • Cat. No.:
    734-3632
  • Storage temperature:
    15...25 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.