ContactUsLinkComponent
pH Sensor, Non-glass, Memosens CPS97E
pH Sensor, Non-glass, Memosens CPS97E
Supplier:  Endress+Hauser
Certificates
pH Sensor, Non-glass, Memosens CPS97E
Supplier:  Endress+Hauser
Specifications
  • Catalog No:
  • ENDRCPS97E-AA8BHH9
  • ENDRCPS97E-AA8BHH2
  • Connection:
  • Inductive, contactless connection head with Memosens 2.0 technology
  • Inductive, contactless connection head with Memosens 2.0 technology
  • pH range:
  • 0 - 14
  • 0 - 14
  • Sensor type:
  • Temperature sensor, Pt1000
  • Temperature sensor, Pt1000
  • Shaft length:
  • 120, 225, 360 and 425 mm
  • 120, 225, 360 and 425 mm
  • Ø:
  • 12 mm
  • 12 mm